Please login to your online account to display your discounted pricing

Bismuth oxychloride, 99.99%, (trace metal basis), ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  212302500

 View more versions of this product

Catalog No. AC212302500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Bismuth (Bi) 79.5 to 80.5 %
Packaging Glass bottle
Total Trace Metal Impurities 100ppm max.
Color White to pink-beige to gray
Quantity 250g
Chemical Name or Material Bismuth(III) oxychloride
CAS 7787-59-9
Purity Grade (trace metal basis)
Physical Form White to pinkish-beige to grey powder
Assay Percent Range 99.99%
Molecular Formula BiClO
Linear Formula O=BiCl
Formula Weight 260.43g/mol
MDL Number MFCD00010895
Merck Index 15, 1274
Synonym Bismuthyl chloride
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
Health Hazard 3 GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio

Specifications

Chemical Name or Material Bismuth(III) oxychloride
CAS 7787-59-9
Purity Grade (trace metal basis)
Assay Percent Range 99.99%
Molecular Formula BiClO
Linear Formula O=BiCl
Formula Weight 260.43g/mol
MDL Number MFCD00010895
Synonym Bismuthyl chloride

For ultra trace analysis

Accessories

Warning

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio

GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.

SDS & Safety Documents