Please login to your online account to display your discounted pricing

Bismuth oxychloride, 99.99%, (trace metal basis), ACROS Organics™

$67.52 - $67.52

Chemical Identifiers

Chemical Name or Material Bismuth(III) oxychloride
CAS 7787-59-9
Purity Grade Trace Metal Basis
Assay Percent Range 99.99%
Molecular Formula BiClO
Linear Formula O=BiCl
Formula Weight 260.43
MDL Number MFCD00010895
Synonym Bismuthyl chloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC212300500

  SDS
thermo fisher scientific
AC212300500
Glass bottle 50g Each for $67.52
Description & Specifications

Specifications

Color White to pinkish-beige to gray
Formula Weight 260.43
Purity Grade Trace Metal Basis
Physical Form Powder
Quantity 50g
Merck Index 15, 1274
Assay Percent Range 99.99%
Molecular Formula BiClO
CAS 7787-59-9
Chemical Name or Material Bismuth(III) oxychloride
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Causes serious eye irritation. May cause respiratory irritation. Causes skin irritation.
MDL Number MFCD00010895
Bismuth (Bi) 79.5 to 80.5 %
Packaging Glass bottle
Linear Formula O=BiCl
Synonym Bismuthyl chloride
Total Trace Metal Impurities 100ppm max.
Health Hazard 3 GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio

For ultra trace analysis