Please login to your online account to display your discounted pricing

Bismuth Nitrate, 5-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  109204

 View more versions of this product

Catalog No. 02002435

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 485.07
Physical Form Crystals
CAS 10035-06-0
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.83kg
Chemical Name or Material Bismuth Nitrate
Quantity 125g
Name Note 5-Hydrate
Molecular Formula Bi(NO3)3·5H2O

Specifications

Chemical Name or Material Bismuth Nitrate
CAS 10035-06-0
Name Note 5-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Bi(NO3)3·5H2O
Formula Weight 485.07