Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Lead Tin ingot (Rose's metal)

Chemical Identifiers

Chemical Name or Material Bismuth Lead Tin ingot (Rose's metal)
MDL Number MFCD00804027
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4051622

  SDS
alfa aesar™
4051622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4051636

  SDS
alfa aesar™
4051636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Alloy
MDL Number MFCD00804027
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn; 50:25:25 wt%
TSCA TSCA
Quantity 100g
Assay Percent Notes (Rose's metal)
Health Hazard 1 Danger
Melting Point 95° to 96°C
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bismuth Lead Tin ingot (Rose's metal)
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.