Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Lead Tin Cadmium eutectic ingot, 99.9% (metals basis)

Low melting application: fuses, etc

Chemical Identifiers

Chemical Name or Material Bismuth Lead Tin Cadmium eutectic ingot
MDL Number MFCD00801077
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40951P2

  SDS
alfa aesar™
40951P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40951Q4

  SDS
alfa aesar™
40951Q4
0.25 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:Sn:Cd; 50.0:26.7:13.3:10.0 wt%
Melting Point 70°C
Chemical Name or Material Bismuth Lead Tin Cadmium eutectic ingot
TSCA TSCA
Assay 99.9%
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1 lb.
Physical Form Ingot
Health Hazard 2 GHS H Statement
H350-H360-H373-H302-H332
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 1 Danger
MDL Number MFCD00801077
Assay Percent Notes (metals basis)
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H360-H373-H302-H332
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.