Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Lead Indium Tin Cadmium eutectic ingot, 99.9% (metals basis)

Low melting application: fuses, etc

Chemical Identifiers

Chemical Name or Material Bismuth Lead Indium Tin Cadmium eutectic ingot
MDL Number MFCD00801081
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40950P2

  SDS
alfa aesar™
40950P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40950Q4

  SDS
alfa aesar™
40950Q4
0.25 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 2 GHS H Statement
H350-H360-H373-H302-H312-H332
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Assay Percent Notes (metals basis)
Quantity 1 lb.
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb:In:Sn:Cd; 44.7:22.6:19.1:8.3:5.3 wt%
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Assay 99.9%
MDL Number MFCD00801081
Chemical Name or Material Bismuth Lead Indium Tin Cadmium eutectic ingot
Health Hazard 1 Danger
Melting Point 47.2°C
Physical Form Ingot
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H360-H373-H302-H312-H332
May cause cancer.
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.