missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Lead eutectic wire, 0.79mm (0.031 in.) dia.

Manufacturer:  Alfa Aesar™ 44238Q4

 View more versions of this product

Catalog No. AA44238Q4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Bi:Pb; 55.5:44.5 wt%
0.25 lb.
MFCD00801141
Wire
Bismuth Lead eutectic wire
0.79mm (0.031 in.) dia.
Insoluble in water.
124°C
Documents