Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Lead eutectic ingot, 99.9% (metals basis)

Low melting application: fuses, etc

Chemical Identifiers

Chemical Name or Material Bismuth Lead eutectic ingot
MDL Number MFCD00801141
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40949P2

  SDS
alfa aesar™
40949P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40949Q4

  SDS
alfa aesar™
40949Q4
0.25 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 1 Danger
Assay Percent Notes (metals basis)
Quantity 1 lb.
TSCA TSCA
Assay 99.9%
Melting Point 124°C
Physical Form Ingot
MDL Number MFCD00801141
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Avg. Mol. Wt. or Mol. Wt. Range Bi:Pb; 55.5:44.5 wt%
Health Hazard 3 GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bismuth Lead eutectic ingot
Safety and Handling

Danger

GHS P Statement
P273-P201-P308+P313-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.