Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth Indium Lead Tin eutectic ingot, alloy 136, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Bismuth Indium Lead Tin eutectic ingot
Name Note Alloy 136
MDL Number MFCD00210606
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1814322

  SDS
alfa aesar™
1814322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

TSCA TSCA
Assay 99.9%
Avg. Mol. Wt. or Mol. Wt. Range Bi:In:Pb:Sn; 49:21:18:12 wt%
Odor Odorless
Health Hazard 2 GHS H Statement
H360-H373-H201
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.
Physical Form Ingot
Health Hazard 1 Danger
Quantity 100g
MDL Number MFCD00210606
Chemical Name or Material Bismuth Indium Lead Tin eutectic ingot
Name Note Alloy 136
Health Hazard 3 GHS P Statement
P260-P281-P308+P313-P314-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Get medical advice/attention if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 58°C
Assay Percent Notes (metals basis)
Safety and Handling

Danger

GHS P Statement
P260-P281-P308+P313-P314-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Get medical advice/attention if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H201
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Explosive
mass explosion hazard.