missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) tert-pentyloxide, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04120303

 View more versions of this product

Catalog No. AA4120303



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bismuth(III) tert-pentyloxide
180°C (decomposition)
63°C (at 10-4mmHg)(sublimation)
MFCD00151666
UN3181
470.41
Powder
124687-44-1
Gray
Bi(OC5H11)3
1 g
Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide
≥97%

Chemical Identifiers

124687-44-1
MFCD00151666
Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only