Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) tert-pentyloxide, 97+%

Chemical Identifiers

Chemical Name or Material Bismuth(III) tert-pentyloxide
CAS 124687-44-1
Linear Formula Bi(OC5H11)3
Formula Weight 470.41
MDL Number MFCD00151666
UN Number UN3181
Synonym Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4120303

  SDS
alfa aesar™
4120303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4120306

  SDS
alfa aesar™
4120306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 63°C (at 10-4mmHg) (subl.)
Formula Weight 470.41
Melting Point 180°C (dec.)
Chemical Name or Material Bismuth(III) tert-pentyloxide
MDL Number MFCD00151666
CAS 124687-44-1
Physical Form Powder
Health Hazard 3 GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
UN Number UN3181
Color Dark gray
Linear Formula Bi(OC5H11)3
Health Hazard 1 Warning
Synonym Bismuth(III) tert-amylate; Bismuth(III) tert-pentoxide
Quantity 1g
Assay ≥97%
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.