Learn More
Bismuth(III) telluride, Vacuum Deposition Grade, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01451918
Description
Bismuth(III) telluride and its alloys (antimony or selenium) posses excellent semiconductor activity, which is used in thermoelectric industry for cooling and energy generation. It increases the speed of microchips and acts as topological insulators in spintronics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth(III) telluride | |
Gray | |
Pieces | |
(metals basis) | |
UN3284 | |
800.76 | |
Odorless |
573°C | |
1304-82-1 | |
50 g | |
MFCD00014201 | |
Insoluble in water. | |
99.999% | |
Moisture sensitive |
Chemical Identifiers
1304-82-1 | |
MFCD00014201 |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TELLURIUM COMPOUND, N.O.S.
EINECSNumber : 215-135-2
RTECSNumber : EB3110000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only