Please login to your online account to display your discounted pricing

Bismuth(III) telluride, Gray powder, 99.98% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Bismuth(III) telluride
CAS 1304-82-1
Physical Form Powder
Assay Percent Range 99.98% (Metals basis)
Molecular Formula Bi2Te3
Formula Weight 800.76
MDL Number MFCD00014201
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4407706

 
alfa aesar
4407706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4407714

 
alfa aesar
4407714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4407722

 
alfa aesar
4407722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 800.76
Quantity 5g
Color Gray
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Bismuth(III) telluride
EINECS Number 215-135-2
Odor Odorless
Assay Percent Range 99.98% (Metals basis)
Molecular Formula Bi2Te3
MDL Number MFCD00014201
CAS 1304-82-1
Health Hazard 1 H331-H302-H312-H315-H319-H335: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Safety and Handling

H331-H302-H312-H315-H319-H335: Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.