Please login to your online account to display your discounted pricing

Bismuth(III) sulfide, Gray lump, 99.999% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Bismuth(III) sulfide
CAS 1345-07-9
Physical Form Lump
Assay Percent Range 99.999% (Metals basis)
Molecular Formula Bi2S3
Formula Weight 514.15
MDL Number MFCD00014204
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4546318

 
alfa aesar
4546318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4546330

 
alfa aesar
4546330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Lump
Formula Weight 514.15
Quantity 50g
Color Gray
Chemical Name or Material Bismuth(III) sulfide
EINECS Number 215-716-0
Molecular Formula Bi2S3
MDL Number MFCD00014204
CAS 1345-07-9
Health Hazard 1 H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.
Density 7.7g/cm3
Assay Percent Range 99.999% (Metals basis)
Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Safety and Handling

H315-H319-H335-H303: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May be harmful if swallowed.

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.