Learn More
Bismuth(III) sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03583418
Description
Bismuth(III) sulfide is used as a starting material to produce other bismuth compounds. It is used in glass industry as a colorant, electronic ceramics, scintillator BGO and in superconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth(III) sulfide | |
1345-07-9 | |
50 g | |
Bi2S3 | |
Insoluble in water. Soluble in acids. | |
[S--].[S--].[S--].[Bi+3].[Bi+3] | |
514.14 | |
99.9% |
Black-Brown | |
Crystals and/or Chunks | |
(metals basis) | |
MFCD00014204 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
dibismuth(3+) trisulfanediide | |
514.15 | |
7.7 g/mL |
Chemical Identifiers
1345-07-9 | |
514.14 | |
YNRGZHRFBQOYPP-UHFFFAOYSA-N | |
[S--].[S--].[S--].[Bi+3].[Bi+3] |
Bi2S3 | |
MFCD00014204 | |
dibismuth(3+) trisulfanediide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 215-716-0
RTECSNumber : EB3105000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only