missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth(III) phosphate, 99%, Thermo Scientific Chemicals
$178.96 - $657.82
Chemical Identifiers
CAS | 10049-01-1 |
---|---|
Molecular Formula | BiH3O4P |
Molecular Weight (g/mol) | 306.97 |
MDL Number | MFCD00049721 |
InChI Key | SFOQXWSZZPWNCL-UHFFFAOYSA-N |
Synonym | bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate, |
PubChem CID | 9839429 |
SMILES | [Bi+3].OP(O)(O)=O |
Description
Bismuth(III) phosphate is used in treatment of renal cysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10049-01-1 | |
306.97 | |
SFOQXWSZZPWNCL-UHFFFAOYSA-N | |
9839429 |
BiH3O4P | |
MFCD00049721 | |
bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate, | |
[Bi+3].OP(O)(O)=O |
Specifications
(decomposition) | |
Powder | |
BiH3O4P | |
14,1275 | |
Insoluble in water and alkali; Soluble in hydrochloric acid | |
[Bi+3].OP(O)(O)=O | |
306.97 | |
303.95 | |
Odorless | |
Bismuth(III) phosphate |
10049-01-1 | |
50 g | |
MFCD00049721 | |
bismuth phosphate, bismuth iii phosphate, bismugel, phosphoric acid, bismuth 3+ salt 1:1, bismuth phosphate mi, bismuth phosphate who-dd, acmc-20allc, bismuth orthophosphate, bi.po4, bismuth phosphate, | |
SFOQXWSZZPWNCL-UHFFFAOYSA-N | |
bismuth;phosphate | |
9839429 | |
99% | |
6.32 g/mL |
Safety and Handling
EINECSNumber : 233-161-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only