missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) oxide, Puratronic™, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 1065818

 View more versions of this product

Catalog No. AA1065818



Specifications

Specifications

Bismuth(III) oxide
(metals basis)
14,1273
Powder
Puratronic™
−20 Mesh
Yellow
50g
1304-76-3
MFCD00003462
465.96
99.9995%
8.9g/mL
~1890°C
817°C

Chemical Identifiers

1304-76-3
MFCD00003462
Safety and Handling

Safety and Handling

RTECSNumber : EB2984460

TSCA : TSCA

Documents