missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) oxide, nanopowder, 99.9%

Manufacturer:  Alfa Aesar™ 4631414

 View more versions of this product

Catalog No. AA4631414



Specifications

Specifications

Bismuth (III) oxide
Bi2O3
14,1273
Nanopowder
8.9g/mL
3.8 to 5.5m2/g
Yellow
25g
1304-76-3
MFCD00003462
465.96
99.9%
80 to 200nm APS
~1890°C
817°C

Chemical Identifiers

1304-76-3
Bi2O3
MFCD00003462
Safety and Handling

Safety and Handling

RTECSNumber : EB2984460

TSCA : TSCA

SDS Documents