missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) oxide, NanoArc™ BI-7300, 99.5+%

Manufacturer:  Alfa Aesar™ 4558214

 View more versions of this product

Catalog No. AA4558214



Description

Description

  • X-ray opaque particle
  • NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties
Specifications

Specifications

Bismuth (III) oxide NanoArc™ BI-7300
Bi2O3
14,1273
Powder
8.9g/mL
38nm APS
~1890°C
817°C
1304-76-3
MFCD00003462
465.96
≥99.5%
Odorless
18m2/g
Yellow
25g

Chemical Identifiers

1304-76-3
Bi2O3
MFCD00003462
Safety and Handling

Safety and Handling

RTECSNumber : EB2984460

TSCA : TSCA

SDS Documents