Learn More
Bismuth(III) nitrate pentahydrate, 99.999%, (trace metal basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 315151000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
80.0°C | |
30.0°C | |
White | |
100 g | |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
bismuth;trinitrate;pentahydrate | |
160911 | |
99.999% | |
Glass bottle |
Bismuth(III) nitrate pentahydrate | |
10035-06-0 | |
Crystals | |
99.999% | |
Bi(NO3)3·5H2O | |
15, 1271 | |
Solubility in water: may decompose | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
485.07 | |
485.07 | |
Trace Metal Basis |
Chemical Identifiers
10035-06-0 | |
485.07 | |
FBXVOTBTGXARNA-UHFFFAOYSA-N | |
160911 | |
O.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
BiH10N3O14 | |
MFCD00149157 | |
bismuth nitrate pentahydrate, bismuth trinitrate pentahydrate, bismuth nitrate, hydrate 1:5, bismuth iii nitrate pentahydrate, unii-fo2icy167b, fo2icy167b, nitric acid, bismuth 3+ salt, pentahydrate, acmc-20alle, bismuthnitratepentahydrate | |
bismuth;trinitrate;pentahydrate |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only