missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) nitrate pentahydrate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 402615000

 View more versions of this product

Catalog No. AC402615000


 

Specifications Description & Specifications

Specifications

≥97.5%
485.07
White
15, 1271
10ppm max.
10ppm max.
10035-06-0
Bismuth(III) nitrate pentahydrate
98%
0.005% max. (in HNO3)
Plastic bottle
10ppm max.
Crystals or Crystalline Powder
500g
For analysis
20ppm max.
10ppm max.
Bi(NO3)3·5H2O
98%
MFCD00149157
BiN3O9·5H2O
20ppm max.
50ppm max.

Chemical Identifiers

10035-06-0
BiN3O9·5H2O
485.07
Bismuth(III) nitrate pentahydrate, 98%
For analysis
98%
Bi(NO3)3·5H2O
MFCD00149157
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : EB2984430