Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)

Chemical Identifiers

Chemical Name or Material Bismuth(III) nitrate oxide
CAS 10361-46-3
Purity Grade Puratronic™
Formula Weight 286.99
MDL Number MFCD00064844
UN Number UN1477
Synonym Bismuth oxynitrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1112209

  SDS
alfa aesar™
1112209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1112218

  SDS
alfa aesar™
1112218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1112230

  SDS
alfa aesar™
1112230
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 286.99
Health Hazard 2 GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99.998%
Synonym Bismuth oxynitrate
UN Number UN1477
Quantity 10g
Chemical Name or Material Bismuth(III) nitrate oxide
Assay Percent Notes (metals basis)
Physical Form Powder
Health Hazard 3 GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
Purity Grade Puratronic™
MDL Number MFCD00064844
CAS 10361-46-3
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.