missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) nitrate oxide, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1112230

 View more versions of this product

Catalog No. AA1112230



Specifications

Specifications

Bismuth(III) nitrate oxide
(metals basis)
UN1477
286.99
99.998%
250g
10361-46-3
MFCD00064844
Bismuth oxynitrate
Powder
Puratronic™

Chemical Identifiers

10361-46-3
MFCD00064844
Bismuth oxynitrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

TSCA : TSCA

Documents