missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) fluoride, anhydrous, 99%

Manufacturing of bismuth pentafluoride

Manufacturer:  Alfa Aesar™ 1307514

 View more versions of this product

Catalog No. AA1307514



Specifications

Specifications

Bismuth(III) fluoride
7787-61-3
MFCD00010893
14,1265
Practically insoluble in water; Soluble in HF
F[Bi](F)F
265.976
265.98
99%
1.74
727°C
Anhydrous
BiF3
UN3260
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii fluoride, bismuth iii fluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
BRCWHGIUHLWZBK-UHFFFAOYSA-K
trifluorobismuthane
82233
Powder
Odorless
Hygroscopic
25g

Chemical Identifiers

7787-61-3
265.976
BRCWHGIUHLWZBK-UHFFFAOYSA-K
82233
F[Bi](F)F
BiF3
MFCD00010893
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii fluoride, bismuth iii fluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
trifluorobismuthane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents