Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) fluoride, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Bismuth(III) fluoride
CAS 7787-61-3
Formula Weight 265.98g/mol
MDL Number MFCD00010893
UN Number UN3260
Merck Index 14,1265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1184414

 
alfa aesar™
11844-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1184422

 
alfa aesar™
11844-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Mesh Size −200 Mesh
Odor Odorless
Quantity 25g
Refractive Index 1.74
Sensitivity Hygroscopic
Melting Point 727°C
Quantity 25g
Chemical Name or Material Bismuth(III) fluoride
CAS 7787-61-3
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
Formula Weight 265.98g/mol
MDL Number MFCD00010893
UN Number UN3260
Merck Index 14,1265
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
I

GHS H Statement
H314-H319
Causes severe skin burns and eye damage.
Causes serious eye irritation.