missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) chloride, White powder, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1711514

 View more versions of this product

Catalog No. AA1711514



Specifications

Specifications

Bismuth(III) chloride
99.99% (Metals basis)
MFCD00003461
Powder
White
7790-78-5
BiCl3
315.34
Irritating
25g

Chemical Identifiers

7790-78-5
BiCl3
MFCD00003461
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 232-123-2

SDS Documents