missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) chloride oxide, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 1713122

 View more versions of this product

Catalog No. AA1713122



Specifications

Specifications

Bismuth(III) chloride oxide
99.95%
BiClHO
MFCD00010895
BWOROQSFKKODDR-UHFFFAOYSA-N
oxobismuth;hydrochloride
6328152
Powder
Odorless
Brown
7787-59-9
(metals basis)
BiOCl
14,1263
O=[Bi].Cl
261.437
260.43
7.72g/mL
2.15
100g

Chemical Identifiers

7787-59-9
261.437
BWOROQSFKKODDR-UHFFFAOYSA-N
oxobismuth;hydrochloride
BiClHO
MFCD00010895
6328152
O=[Bi].Cl
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EB2700000

TSCA : TSCA

SDS Documents