missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth(III) chloride, anhydrous, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 8948514

 View more versions of this product

Catalog No. AA8948514



Specifications

Specifications

Bismuth (III) chloride
7787-60-2
BiCl3
UN3260
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth iii chloride, bismuth lll chloride, bismuth iii chloride
Cl[Bi](Cl)Cl
315.33
315.34
99.999%
Hygroscopic
White
25g
Anhydrous
(metals basis)
MFCD00003461
14,1262
JHXKRIRFYBPWGE-UHFFFAOYSA-K
trichlorobismuthane
24591
Crystalline
4.75g/mL
~430°C (sublimation)
230°C to 232°C

Chemical Identifiers

7787-60-2
315.33
JHXKRIRFYBPWGE-UHFFFAOYSA-K
24591
Cl[Bi](Cl)Cl
BiCl3
MFCD00003461
bismuth trichloride, bismuth chloride, trichlorobismuthine, trichlorobismuth, bismuth iii chloride, bismuthine, trichloro, trichlorobismute, bismuth iii chloride, bismuth lll chloride, bismuth iii chloride
trichlorobismuthane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : EB2690000

TSCA : TSCA

SDS Documents