Please login to your online account to display your discounted pricing

Bismuth(III) chloride, +98%, anhydrous, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC208835000

 View more versions of this product

Catalog No. AC208835000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder and/or Chunks
Formula Weight 315.33
Quantity 500g
Color White to beige or gray
Density 4.7500g/mL
Sublimation Point 430 °C
Merck Index 15, 1264
Fieser 07,24
Assay Percent Range 98+%
MDL Number MFCD00003461
Packaging Glass bottle
Linear Formula BiCl3
Name Note Anhydrous
Molecular Formula BiCl3
Chemical Name or Material Bismuth(III) chloride
CAS 7787-60-2
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Bismuth(III) chloride
Name Note Anhydrous
CAS 7787-60-2
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula BiCl3
Linear Formula BiCl3
Formula Weight 315.33
MDL Number MFCD00003461
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents