missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth(III) bromide, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1711222

 View more versions of this product

Catalog No. AA1711222



Specifications

Specifications

Bismuth(III) bromide
99% (metals basis)
MFCD00014195
TXKAQZRUJUNDHI-UHFFFAOYSA-K
tribromobismuthane
82232
Powder
100g
7787-58-8
BiBr3
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo, bismuth iii bromide
Br[Bi](Br)Br
448.692
448.71
5.72

Chemical Identifiers

7787-58-8
448.692
TXKAQZRUJUNDHI-UHFFFAOYSA-K
82232
Br[Bi](Br)Br
BiBr3
MFCD00014195
bismuth tribromide, bismuth iii bromide, bismuth bromide, bismuthine, tribromo, tribromobismuthine, unii-dt59198t2x, bismuth ii bromide, tribromobismuth, bismuthine,tribromo, bismuth iii bromide
tribromobismuthane
Safety and Handling

Safety and Handling

EINECSNumber : 232-121-1

SDS Documents