missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth(III) acetate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01757430
Description
Bismuth(III) acetate is used in biology, printing, in photographic films, textiles, and also in proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth(III) acetate | |
White | |
Crystalline | |
C6H9BiO6 | |
MFCD00015629 | |
Soluble in water. | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3] | |
386.112 | |
386.12 | |
Hygroscopic |
(decomposition) | |
22306-37-2 | |
250 g | |
Bi(OOCCH3)3 | |
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g | |
WKLWZEWIYUTZNJ-UHFFFAOYSA-K | |
bismuth;triacetate | |
31132 | |
99% |
Chemical Identifiers
22306-37-2 | |
386.112 | |
WKLWZEWIYUTZNJ-UHFFFAOYSA-K | |
31132 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Bi+3] |
C6H9BiO6 | |
MFCD00015629 | |
bismuth acetate, bismuth triacetate, unii-8aja86y692, acetic acid, bismuth 3+ salt, acmc-20allk, bismuth iii ; acetate, bismuth 3+ triacetate, trisacetic acid bismuth salt, bismuth 3+ ion tris acetate ion, bismuth iii acetate 50g | |
bismuth;triacetate |
Safety and Handling
EINECSNumber : 244-904-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only