Please login to your online account to display your discounted pricing

Bismuth hydroxide, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC393292500

 View more versions of this product

Catalog No. AC393292500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 260
Quantity 250g
Color White to cream
Merck Index 15, 1267
Identification Positive
Packaging Glass bottle
Linear Formula Bi(OH)3
Bismuth (Bi) 80.0% min. (on dry basis)
Molecular Formula H3BiO3
Chemical Name or Material Bismuth hydroxide
MDL Number MFCD00046188
Loss on Drying 1.0% max.
CAS 10361-43-0

Specifications

Chemical Name or Material Bismuth hydroxide
CAS 10361-43-0
Molecular Formula H3BiO3
Linear Formula Bi(OH)3
Formula Weight 260
MDL Number MFCD00046188