Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth hexafluoro-2,4-pentanedionate, 97+%

$275.74 - $275.74

Chemical Identifiers

Chemical Name or Material Bismuth hexafluoro-2,4-pentanedionate
CAS 142617-56-9
Formula Weight 830.14
MDL Number MFCD00270924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4120106

  SDS
alfa aesar™
4120106
5g Each for $275.74
Description & Specifications

Specifications

Formula Weight 830.14
Melting Point ≈188°C
Quantity 5g
Health Hazard 1 Warning
Assay ≥97%
CAS 142617-56-9
Physical Form Powder
Chemical Name or Material Bismuth hexafluoro-2,4-pentanedionate
MDL Number MFCD00270924
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 110°C (0.05mmHg) (subl.)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.