Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth aluminum oxide hydrate, 95.5%

Chemical Identifiers

Chemical Name or Material Bismuth aluminum oxide hydrate
Formula Weight 771.84 (Anhydrous)
MDL Number MFCD00798526
Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4039318

  SDS
alfa aesar™
4039318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4039330

  SDS
alfa aesar™
4039330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40393A1

  SDS
alfa aesar™
40393A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 771.84 (Anhydrous)
Synonym Aluminum bismuth oxide hydrate; Bismuth aluminate hydrate
Solubility Information Insoluble in water; Soluble in nitric acid; Typical tap density 0;69g/cm3
Chemical Name or Material Bismuth aluminum oxide hydrate
MDL Number MFCD00798526
Physical Form Amorphous Powder
Assay 95.5%
Quantity 50g
Particle Size typically 2 Micron max