missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Bi), LabChem™

Manufacturer:  LabChem LC116707

 View more versions of this product

Catalog No. LC116707



Specifications

Specifications

Bismuth AA Standard
10035-06-0,7697-37-2,7732-18-5
BiH10N3O14
1000ppm ±10ppm (1mL = 1mg Bi)
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth;trinitrate;pentahydrate
160911
Liquid
Certified
Soluble in water
0.99cSt
Colorless
1000ppm (1mL = 1mg Bi)
95.92,3.85,0.23
Bi(NO2)3·5H2O
UN2031
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Bi+3]
485.067
485.07
Passes Test
Poly Bottle
Traceable to NIST
1.02g/mL
125mL

Chemical Identifiers

10035-06-0
485.067
160911
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.[Bi+3]
BiH10N3O14
FBXVOTBTGXARNA-UHFFFAOYSA-N
bismuth;trinitrate;pentahydrate
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions