Please login to your online account to display your discounted pricing

Bismuth AA Standard, 1mL = 1mg Bi (1,000ppm Bi), Ricca Chemical

$55.45 - $55.45

Chemical Identifiers

Chemical Name or Material Bismuth AA Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.92
CAS Max % 5.12
CAS 7440-69-9
CAS Min % 0.1
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ABI1KN-100

 
ricca chemical company
RIABI1KN100
Poly natural 100mL Each for $55.45

ABI1KN-500

 
ricca chemical company
RIABI1KN500
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 100mL
Equivalence 1 mL = 1 mg Bi
Concentration or Composition Notes 1,000ppm Bi
Chemical Name or Material Bismuth AA Standard
CAS 7732-18-5
Packaging Poly natural
CAS 7697-37-2
CAS Max % 5.12
CAS Min % 0.1
CAS Min % 4.92
CAS Max % 97.0
CAS Min % 93.0
CAS Max % 0.1
CAS 7440-69-9