missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth 2-ethylhexanoate, typically 92%, in 2-ethylhexanoic acid

Manufacturer:  Alfa Aesar™ 4550006

 View more versions of this product

Catalog No. AA4550006



Specifications

Specifications

Bismuth 2-ethylhexanoate
In 2-ethylhexanoic acid
C24H48BiO6
2-Ethylhexanoic acid bismuth salt
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]
641.622
638.61
92% (typically)
5g
67874-71-9
MFCD00043803
MWXWNYHYPMRCHN-UHFFFAOYSA-N
bismuth;2-ethylhexanoic acid
131857451
Liquid

Chemical Identifiers

67874-71-9
641.622
MWXWNYHYPMRCHN-UHFFFAOYSA-N
131857451
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]
C24H48BiO6
MFCD00043803
2-Ethylhexanoic acid bismuth salt
bismuth;2-ethylhexanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents