missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth 2-ethylhexanoate acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04550022
Description
Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth 2-ethylhexanoate | |
67874-71-9 | |
MFCD00043803 | |
2-Ethylhexanoic acid bismuth salt | |
MWXWNYHYPMRCHN-UHFFFAOYSA-N | |
bismuth;2-ethylhexanoic acid | |
131857451 | |
92% (typically) |
In 2-ethylhexanoic acid | |
C24H48BiO6 | |
100 g | |
Insoluble in water. | |
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi] | |
641.622 | |
638.61 | |
Liquid |
Chemical Identifiers
67874-71-9 | |
641.622 | |
MWXWNYHYPMRCHN-UHFFFAOYSA-N | |
131857451 | |
CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi] |
C24H48BiO6 | |
MFCD00043803 | |
2-Ethylhexanoic acid bismuth salt | |
bismuth;2-ethylhexanoic acid |
Safety and Handling
GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
P201-P202-P281-P308+P313-P501c
H361d
EINECSNumber : 267-499-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only