Please login to your online account to display your discounted pricing

Alfa Aesar™ Bismuth 2-ethylhexanoate, 99.99+%, 70% w/v in xylene

Chemical Identifiers

Chemical Name or Material Bismuth 2-ethylhexanoate
CAS 67874-71-9
Formula Weight 638.61
MDL Number MFCD00043803
UN Number UN1307
Synonym 2-Ethylhexanoic acid bismuth salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4152106

  SDS
alfa aesar™
4152106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4152114

  SDS
alfa aesar™
4152114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4152122

  SDS
alfa aesar™
4152122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 638.61
Quantity 5g
Physical Form Liquid
Assay ≥99.99%
CAS 67874-71-9
Synonym 2-Ethylhexanoic acid bismuth salt
Chemical Name or Material Bismuth 2-ethylhexanoate
Concentration 70% w/v in xylene
TSCA TSCA
Health Hazard 3 GHS P Statement
P280h-P262
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.
UN Number UN1307
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H361-H312-H332-H315
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
MDL Number MFCD00043803
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: XYLENES
Safety and Handling

Warning

GHS P Statement
P280h-P262
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.

GHS H Statement
H226-H361-H312-H332-H315
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.