missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth, AAS standard solution, Specpure™ Bi 1000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 088054AP
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bismuth | |
500 mL | |
Bi | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
bismuth | |
5359367 | |
AAS Standard | |
17mm Hg at 20°C |
7440-69-9,7732-18-5 | |
Colorless | |
MFCD00134033 | |
[Bi] | |
208.98 | |
CHEBI:33301 | |
Nitric acid: 5%; Bismuth: 0.1%; Water: 94.9% | |
Liquid |
Chemical Identifiers
7440-69-9 | |
208.98 | |
JCXGWMGPZLAOME-UHFFFAOYSA-N | |
CHEBI:33301 | |
[Bi] |
Bi | |
MFCD00134033 | |
5359367 | |
bismuth |
Safety and Handling
P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-714-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only