missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth, 1000μg/mL, AAS standard solution, Alfa Aesar™ Specpure™

Manufacturer:  Alfa Aesar 88054AE

 View more versions of this product

Catalog No. AA88054AE



Specifications

Specifications

Bismuth
AAS Standard
MFCD00134033
JCXGWMGPZLAOME-UHFFFAOYSA-N
bismuth
5359367
Liquid
Colorless
7440-69-9,7732-18-5,7732-18-5
Bi
Nitric acid: 5%; Bismuth: 0.1%; Water: 94.9%
[Bi]
208.98
CHEBI:33301
17mm Hg at 20°C
100mL

Chemical Identifiers

7440-69-9
208.98
JCXGWMGPZLAOME-UHFFFAOYSA-N
CHEBI:33301
[Bi]
Bi
MFCD00134033
5359367
bismuth
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-714-2

SDS Documents