Please login to your online account to display your discounted pricing

Bisacrylamide, Molecular Grade, Promega™ DFS Item


Quantity 25g

Manufacturer: promega corportaion  V3141

 View more versions of this product

Catalog No. PR-V3141

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 154.2
Purity Grade Molecular Grade
Physical Form Free-Flowing Crystals
Color White
Quantity 25g
Synonym N, N«-Methylenebisacrylamide F.W. 154.20
Recommended Storage Store at 22° to 25°C.
Chemical Name or Material Bisacrylamide

Specifications

Chemical Name or Material Bisacrylamide
Purity Grade Molecular Grade
Physical Form Free-Flowing Crystals
Formula Weight 154.2
Synonym N, N«-Methylenebisacrylamide F.W. 154.20
Accessories

Store at 22° to 25°C.