Please login to your online account to display your discounted pricing

Bisacrylamide, Molecular Grade, Promega™

$39.59 - $120.91

Chemical Identifiers

Chemical Name or Material Bisacrylamide
Purity Grade Molecular Grade
Physical Form Free-Flowing Crystals
Formula Weight 154.2
Synonym N, N«-Methylenebisacrylamide F.W. 154.20
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

PR-V3141

 
promega corportaion
V3141
25g Each for $39.59

PRV3143

 
promega corportaion
V3143
125g Each for $120.91
Description & Specifications

Specifications

Purity Grade Molecular Grade
Physical Form Free-Flowing Crystals
Formula Weight 154.2
Color White
Quantity 25g
Chemical Name or Material Bisacrylamide
Synonym N, N«-Methylenebisacrylamide F.W. 154.20
Recommended Storage Store at 22° to 25°C.
Safety and Handling

Store at 22° to 25°C.