Learn More
Bisacodyl, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6199714
Description
Bisacodyl is used in the inhibition of (Na+K)ATPase, the increase of mucosal PGE2, and the activation of adenyl cyclase in rat intestine studies. It is used as a laxative drug. It is typically prescribed for relief of constipation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in alcohol (slightly), ether (slightly), chloroform, and acetone. Insoluble in water.
Notes
Protect from heat. Incompatible with heat and oxidizing agents. Store at 4°C.
Specifications
Bisacodyl | |
White | |
MFCD00038039 | |
14,1242 | |
Soluble in alcohol (slightly),ether (slightly),chloroform,and acetone. Insoluble in water. | |
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1 | |
361.40 | |
361.4 | |
Crystalline Powder |
603-50-9 | |
C22H19NO4 | |
25 g | |
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual | |
KHOITXIGCFIULA-UHFFFAOYSA-N | |
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate | |
2391 | |
≥98% |
Chemical Identifiers
603-50-9 | |
361.40 | |
KHOITXIGCFIULA-UHFFFAOYSA-N | |
2391 | |
CC(=O)OC1=CC=C(C=C1)C(C1=CC=C(OC(C)=O)C=C1)C1=CC=CC=N1 |
C22H19NO4 | |
MFCD00038039 | |
bisacodyl, dulcolax, bicol, brocalax, durolax, dulcolan, endokolat, fenilaxan, hillcolax, sanvacual | |
4-{[4-(acetyloxy)phenyl](pyridin-2-yl)methyl}phenyl acetate |
Safety and Handling
EINECSNumber : 210-044-4
RTECSNumber : SM8750000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only