Please login to your online account to display your discounted pricing

BIS-Tris Propane, ULTROL™ Grade, Calbiochem™, EMD Millipore

A non to zwitterionic buffer that is useful in the pH 6.3 to 9.5 range

$166.13 - $166.13

Chemical Identifiers

Assay Percent Range >98%
Formula Weight 282.4
Buffers non-zwitterionic buffer
CAS 64431-96-5
Chemical Name or Material BIS-Tris Propane
Molecular Formula C11H26N2O6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

394111100GM

 
emd millipore corporation
394111100GM
Each for $166.13
Description & Specifications

Specifications

Formula Weight 282.4
Quantity 100g
Chemical Name or Material BIS-Tris Propane
Buffers non-zwitterionic buffer
Purity Grade Notes by titration
Assay Percent Range >98%
Molecular Formula C11H26N2O6
CAS 64431-96-5
Synonym 1,3-bis[tris(Hydroxymethyl)methylamino]propane
Solubility Information Easily soluble in cold water
Purity ULTROL™ Grade
Recommended Storage 15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Safety and Handling

15°C to 30°C (Do not freeze); Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).