missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ BIS-TRIS propane, 0.2M buffer soln., pH 7.5

$84.96 - $133.08

Chemical Identifiers

CAS 64431-96-5
Molecular Formula C11H26N2O6
Molecular Weight (g/mol) 282.337
MDL Number MFCD00004689
InChI Key HHKZCCWKTZRCCL-UHFFFAOYSA-N
PubChem CID 125132
ChEBI CHEBI:40947
IUPAC Name 2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
SMILES C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61292AE View Documents Alfa Aesar™
J61292AE
100mL Each for $84.96
 
AAJ61292AK View Documents Alfa Aesar™
J61292AK
250mL Each for $133.08
 
Specifications

Chemical Identifiers

64431-96-5
282.337
HHKZCCWKTZRCCL-UHFFFAOYSA-N
CHEBI:40947
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
C11H26N2O6
MFCD00004689
125132
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol

Specifications

64431-96-5
MFCD00004689
HHKZCCWKTZRCCL-UHFFFAOYSA-N
2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
125132
Liquid
100mL
C11H26N2O6
0.2M buffer soln.
C(CNC(CO)(CO)CO)CNC(CO)(CO)CO
282.337
CHEBI:40947
7.5
BIS-TRIS propane buffer soln.