missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
bis-Tris, ≥99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210103890
Description
- Zwitterionic buffer
- Buffering agent
- White Powder
Specifications
bis-Tris | |
>99% | |
zwitterionic buffer | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol | |
81462 | |
209.2 | |
White | |
White Powder |
6976-37-0 | |
C8H19NO5 | |
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane | |
C(CO)N(CCO)C(CO)(CO)CO | |
209.242 | |
CHEBI:41250 | |
Crystalline Powder | |
500g |
Chemical Identifiers
6976-37-0 | |
209.242 | |
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 2-Bis(2-hydroxyethyl)amino-2-(hydroxymethyl)-1,3-propanediol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane | |
CHEBI:41250 | |
C(CO)N(CCO)C(CO)(CO)CO |
C8H19NO5 | |
OWMVSZAMULFTJU-UHFFFAOYSA-N | |
81462 | |
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol |