Please login to your online account to display your discounted pricing

Alfa Aesar™ BIS-TRIS, 0.2M buffer soln., pH 7.4, low endotoxin

Chemical Identifiers

Chemical Name or Material BIS-TRIS
CAS 6976-37-0
Name Note low endotoxin
MDL Number MFCD00002853
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67430AE

 
alfa aesar™
J67430AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67430AK

 
alfa aesar™
J67430AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material BIS-TRIS
MDL Number MFCD00002853
Physical Form Liquid
Name Note low endotoxin
Concentration 0.2M buffer soln.
TSCA TSCA
Storage Note 1 2° to 8°C
Color Colorless
Quantity 100mL
CAS 6976-37-0
pH 7.4
Safety and Handling

2° to 8°C