missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(triphenyltin) oxide, 98%, ACROS Organics™

Chemical Identifiers

CAS 1262-21-1
Molecular Formula C36H30OSn2
Molecular Weight (g/mol) 716.055
MDL Number MFCD00003005
InChI Key MUHFQLVFMFDJOK-UHFFFAOYSA-N
Synonym bis triphenyl tin oxide, bis triphenyltin oxide, bis triphenylzinn-oxid, distannoxane, hexaphenyl, fentin oxide, fentin-oxide, muhfqlvfmfdjok-uhfffaoysa, triphenyl triphenylstannyl oxy stannane, triphenyl triphenylstannyloxy stannane
PubChem CID 16683296
IUPAC Name triphenyl(triphenylstannyloxy)stannane
SMILES C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(C3=CC=CC=C3)O[Sn](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC151130250
SDS View Documents
Thermo Fisher Scientific
151130250
25g Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1262-21-1
716.055
MUHFQLVFMFDJOK-UHFFFAOYSA-N
16683296
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(C3=CC=CC=C3)O[Sn](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6
C36H30OSn2
MFCD00003005
bis triphenyl tin oxide, bis triphenyltin oxide, bis triphenylzinn-oxid, distannoxane, hexaphenyl, fentin oxide, fentin-oxide, muhfqlvfmfdjok-uhfffaoysa, triphenyl triphenylstannyl oxy stannane, triphenyl triphenylstannyloxy stannane
triphenyl(triphenylstannyloxy)stannane

Specifications

Authentic
White
25g
98%
[(C6H5)3Sn]2O
bis triphenyl tin oxide, bis triphenyltin oxide, bis triphenylzinn-oxid, distannoxane, hexaphenyl, fentin oxide, fentin-oxide, muhfqlvfmfdjok-uhfffaoysa, triphenyl triphenylstannyl oxy stannane, triphenyl triphenylstannyloxy stannane
C1=CC=C(C=C1)[Sn](C2=CC=CC=C2)(C3=CC=CC=C3)O[Sn](C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6
716.055
716.02
98%
Glass Bottle
119.0°C to 123.0°C
1262-21-1
C36H30OSn2
MFCD00003005
MUHFQLVFMFDJOK-UHFFFAOYSA-N
triphenyl(triphenylstannyloxy)stannane
16683296
Crystalline Powder and/or Crystals
Bis(triphenyltin) oxide
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
Toxic if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 215-025-4