Please login to your online account to display your discounted pricing

Alfa Aesar™ Bis(triphenyltin) oxide, 98%

Chemical Identifiers

Chemical Name or Material Bis(triphenyltin) oxide
CAS 1262-21-1
Molecular Formula [(C6H5)3Sn]2O
Formula Weight 716.06g/mol
MDL Number MFCD00003005
UN Number UN3146
Beilstein 4166537
Synonym Hexaphenyl distannoxane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2360906

 
alfa aesar™
B23609-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 119° to 123°C
Quantity 5g
Chemical Name or Material Bis(triphenyltin) oxide
CAS 1262-21-1
Assay 98%
Molecular Formula [(C6H5)3Sn]2O
Formula Weight 716.06g/mol
MDL Number MFCD00003005
UN Number UN3146
Beilstein 4166537
Synonym Hexaphenyl distannoxane
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.